Anti-NDUFB10 Mouse Monoclonal Antibody [clone: 4D1]
Toimittaja: OriGene
ORIGCF505170EA 770 EUR
ORIGCF505170
Anti-NDUFB10 Mouse Monoclonal Antibody [clone: 4D1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: NDUFB10
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human