Anti-ALX4 Mouse Monoclonal Antibody [clone: 6B3]
Toimittaja: OriGene
ORIGCF505173EA 749 EUR
ORIGCF505173
Anti-ALX4 Mouse Monoclonal Antibody [clone: 6B3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: ALX4
Clonality: Monoclonal
Clone: 6B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human