Anti-PPIL6 Mouse Monoclonal Antibody [clone: 4C1]

Toimittaja: OriGene
ORIGCF504839EA 749 EUR
ORIGCF504839
Anti-PPIL6 Mouse Monoclonal Antibody [clone: 4C1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PPIL6
Clonality: Monoclonal
Clone: 4C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR