Anti-ZSCAN18 Mouse Monoclonal Antibody [clone: 2D3]
Toimittaja: OriGene
ORIGCF505242EA 770 EUR
ORIGCF505242
Anti-ZSCAN18 Mouse Monoclonal Antibody [clone: 2D3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ZSCAN18
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human