Anti-ZSCAN18 Mouse Monoclonal Antibody [clone: 4B7]

Toimittaja: OriGene
ORIGCF505243EA 770 EUR
ORIGCF505243
Anti-ZSCAN18 Mouse Monoclonal Antibody [clone: 4B7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ZSCAN18
Clonality: Monoclonal
Clone: 4B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR