Anti-F11R Mouse Monoclonal Antibody [clone: 6E11]

Toimittaja: OriGene
ORIGCF506034EA 749 EUR
ORIGCF506034
Anti-F11R Mouse Monoclonal Antibody [clone: 6E11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: F11R
Clonality: Monoclonal
Clone: 6E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR