Anti-FGFR2 Mouse Monoclonal Antibody [clone: 3B3]

Toimittaja: OriGene
ORIGCF502877EA 749 EUR
ORIGCF502877
Anti-FGFR2 Mouse Monoclonal Antibody [clone: 3B3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: FGFR2
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR