Anti-UBE2G2 Mouse Monoclonal Antibody [clone: 1D7]

Toimittaja: OriGene
ORIGCF505258EA 770 EUR
ORIGCF505258
Anti-UBE2G2 Mouse Monoclonal Antibody [clone: 1D7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: UBE2G2
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR