Anti-SRY Mouse Monoclonal Antibody [clone: 2C7]

Toimittaja: OriGene
ORIGCF505290EA 749 EUR
ORIGCF505290
Anti-SRY Mouse Monoclonal Antibody [clone: 2C7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SRY
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR