Anti-NKIRAS1 Mouse Monoclonal Antibody [clone: 1D2]
Toimittaja: OriGene
ORIGCF505298EA 749 EUR
ORIGCF505298
Anti-NKIRAS1 Mouse Monoclonal Antibody [clone: 1D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: NKIRAS1
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human