Anti-HSPBP1 Mouse Monoclonal Antibody [clone: 2C8]

Toimittaja: OriGene
ORIGCF503311EA 770 EUR
ORIGCF503311
Anti-HSPBP1 Mouse Monoclonal Antibody [clone: 2C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HSPBP1
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR