Anti-RNF113B Mouse Monoclonal Antibody [clone: 7A1]

Toimittaja: OriGene
ORIGCF504117EA 749 EUR
ORIGCF504117
Anti-RNF113B Mouse Monoclonal Antibody [clone: 7A1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RNF113B
Clonality: Monoclonal
Clone: 7A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR