Anti-C1orf50 Mouse Monoclonal Antibody [clone: 4D10]
Toimittaja: OriGene
ORIGCF502938EA 770 EUR
ORIGCF502938
Anti-C1orf50 Mouse Monoclonal Antibody [clone: 4D10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: C1orf50
Clonality: Monoclonal
Clone: 4D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human