Anti-NEU2 Mouse Monoclonal Antibody [clone: 4F4]

Toimittaja: OriGene
ORIGCF502942EA 770 EUR
ORIGCF502942
Anti-NEU2 Mouse Monoclonal Antibody [clone: 4F4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NEU2
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR