Anti-LGR5 Mouse Monoclonal Antibody [clone: 3E11]

Toimittaja: OriGene
ORIGCF502950EA 749 EUR
ORIGCF502950
Anti-LGR5 Mouse Monoclonal Antibody [clone: 3E11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LGR5
Clonality: Monoclonal
Clone: 3E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR