Anti-FGF21 Mouse Monoclonal Antibody [clone: 1H4]

Toimittaja: OriGene
ORIGCF502953EA 770 EUR
ORIGCF502953
Anti-FGF21 Mouse Monoclonal Antibody [clone: 1H4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FGF21
Clonality: Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR