Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 1F5]

Toimittaja: OriGene
ORIGCF505335EA 770 EUR
ORIGCF505335
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 1F5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: CCT8L2
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR