Anti-RAB37 Mouse Monoclonal Antibody [clone: 2F8]

Toimittaja: OriGene
ORIGCF505338EA 749 EUR
ORIGCF505338
Anti-RAB37 Mouse Monoclonal Antibody [clone: 2F8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RAB37
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR