Anti-PPIL3 Mouse Monoclonal Antibody [clone: 2E7]

Toimittaja: OriGene
ORIGCF505349EA 749 EUR
ORIGCF505349
Anti-PPIL3 Mouse Monoclonal Antibody [clone: 2E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PPIL3
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR