Anti-DEF6 Mouse Monoclonal Antibody [clone: 1D1]
Toimittaja: OriGene
ORIGCF505352EA 749 EUR
ORIGCF505352
Anti-DEF6 Mouse Monoclonal Antibody [clone: 1D1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:100
Type: Primary
Antigen: DEF6
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human