Anti-IRF6 Mouse Monoclonal Antibody [clone: 1G3]
Toimittaja: OriGene
ORIGCF503382EA 749 EUR
ORIGCF503382
Anti-IRF6 Mouse Monoclonal Antibody [clone: 1G3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: IRF6
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human