Anti-PGAM2 Mouse Monoclonal Antibody [clone: 3B11]

Toimittaja: OriGene
ORIGCF503401EA 749 EUR
ORIGCF503401
Anti-PGAM2 Mouse Monoclonal Antibody [clone: 3B11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PGAM2
Clonality: Monoclonal
Clone: 3B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR