Anti-ANAPC2 Mouse Monoclonal Antibody [clone: 1B12]
Toimittaja: OriGene
ORIGCF503404EA 770 EUR
ORIGCF503404
Anti-ANAPC2 Mouse Monoclonal Antibody [clone: 1B12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100
Type: Primary
Antigen: ANAPC2
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human