Anti-GOLM1 Mouse Monoclonal Antibody [clone: 7B3]

Toimittaja: OriGene
ORIGCF504187EA 749 EUR
ORIGCF504187
Anti-GOLM1 Mouse Monoclonal Antibody [clone: 7B3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GOLM1
Clonality: Monoclonal
Clone: 7B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR