Anti-ENDOG Mouse Monoclonal Antibody [clone: 5F1]

Toimittaja: OriGene
ORIGCF504189EA 770 EUR
ORIGCF504189
Anti-ENDOG Mouse Monoclonal Antibody [clone: 5F1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ENDOG
Clonality: Monoclonal
Clone: 5F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR