Anti-LIMK1 Mouse Monoclonal Antibody [clone: 8B2]

Toimittaja: OriGene
ORIGCF502977EA 749 EUR
ORIGCF502977
Anti-LIMK1 Mouse Monoclonal Antibody [clone: 8B2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LIMK1
Clonality: Monoclonal
Clone: 8B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR