Anti-PDF Mouse Monoclonal Antibody [clone: 3F3]

Toimittaja: OriGene
ORIGCF502988EA 770 EUR
ORIGCF502988
Anti-PDF Mouse Monoclonal Antibody [clone: 3F3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDF
Clonality: Monoclonal
Clone: 3F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR