Anti-ETS2 Mouse Monoclonal Antibody [clone: 2A3]

Toimittaja: OriGene
ORIGCF505391EA 770 EUR
ORIGCF505391
Anti-ETS2 Mouse Monoclonal Antibody [clone: 2A3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ETS2
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR