Anti-CCM2 Mouse Monoclonal Antibody [clone: 2F9]
Toimittaja: OriGene
ORIGCF503414EA 749 EUR
ORIGCF503414
Anti-CCM2 Mouse Monoclonal Antibody [clone: 2F9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: CCM2
Clonality: Monoclonal
Clone: 2F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human