Anti-CYP17A1 Mouse Monoclonal Antibody [clone: 3F11]

Toimittaja: OriGene
ORIGCF503442EA 749 EUR
ORIGCF503442
Anti-CYP17A1 Mouse Monoclonal Antibody [clone: 3F11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP17A1
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR