Anti-ERBB2 Mouse Monoclonal Antibody [clone: 4F10]
Toimittaja: OriGene
ORIGCF503443EA 749 EUR
ORIGCF503443
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 4F10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human