Anti-ASNS Mouse Monoclonal Antibody [clone: 1A10]

Toimittaja: OriGene
ORIGCF503452EA 749 EUR
ORIGCF503452
Anti-ASNS Mouse Monoclonal Antibody [clone: 1A10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ASNS
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR