Anti-EXOSC7 Mouse Monoclonal Antibody [clone: 3B2]

Toimittaja: OriGene
ORIGCF504243EA 749 EUR
ORIGCF504243
Anti-EXOSC7 Mouse Monoclonal Antibody [clone: 3B2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: EXOSC7
Clonality: Monoclonal
Clone: 3B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR