Anti-C1S Mouse Monoclonal Antibody [clone: 2A8]

Toimittaja: OriGene
ORIGCF504247EA 785 EUR
ORIGCF504247
Anti-C1S Mouse Monoclonal Antibody [clone: 2A8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: C1S
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR