Anti-AP2M1 Mouse Monoclonal Antibody [clone: 1C8]

Toimittaja: OriGene
ORIGCF503018EA 749 EUR
ORIGCF503018
Anti-AP2M1 Mouse Monoclonal Antibody [clone: 1C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: AP2M1
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR