Anti-ITFG2 Mouse Monoclonal Antibody [clone: 3A2]

Toimittaja: OriGene
ORIGCF503051EA 749 EUR
ORIGCF503051
Anti-ITFG2 Mouse Monoclonal Antibody [clone: 3A2]
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ITFG2
Clonality: Monoclonal
Clone: 3A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR