Anti-SOCS3 Mouse Monoclonal Antibody [clone: 15F9]

Toimittaja: OriGene
ORIGCF503054EA 749 EUR
ORIGCF503054
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 15F9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 15F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR