Anti-COG6 Mouse Monoclonal Antibody [clone: 1B8]

Toimittaja: OriGene
ORIGCF505418EA 749 EUR
ORIGCF505418
Anti-COG6 Mouse Monoclonal Antibody [clone: 1B8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: COG6
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR