Anti-DCPS Mouse Monoclonal Antibody [clone: 1E6]

Toimittaja: OriGene
ORIGCF505420EA 749 EUR
ORIGCF505420
Anti-DCPS Mouse Monoclonal Antibody [clone: 1E6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: DCPS
Clonality: Monoclonal
Clone: 1E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR