Anti-PCDH7 Mouse Monoclonal Antibody [clone: 2G6]

Toimittaja: OriGene
ORIGCF505452EA 749 EUR
ORIGCF505452
Anti-PCDH7 Mouse Monoclonal Antibody [clone: 2G6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PCDH7
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR