Anti-PDE4B Mouse Monoclonal Antibody [clone: 1D12]

Toimittaja: OriGene
ORIGCF503471EA 749 EUR
ORIGCF503471
Anti-PDE4B Mouse Monoclonal Antibody [clone: 1D12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4B
Clonality: Monoclonal
Clone: 1D12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR