Anti-RFXANK Mouse Monoclonal Antibody [clone: 2G7]

Toimittaja: OriGene
ORIGCF504294EA 749 EUR
ORIGCF504294
Anti-RFXANK Mouse Monoclonal Antibody [clone: 2G7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RFXANK
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR