Anti-LENG1 Mouse Monoclonal Antibody [clone: 1G10]

Toimittaja: OriGene
ORIGCF503083EA 770 EUR
ORIGCF503083
Anti-LENG1 Mouse Monoclonal Antibody [clone: 1G10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LENG1
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR