Anti-TAGLN Mouse Monoclonal Antibody [clone: 8C8]
Toimittaja: OriGene
ORIGCF503092EA 749 EUR
ORIGCF503092
Anti-TAGLN Mouse Monoclonal Antibody [clone: 8C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: TAGLN
Clonality: Monoclonal
Clone: 8C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse