Anti-SMS Mouse Monoclonal Antibody [clone: 2H1]

Toimittaja: OriGene
ORIGCF503101EA 749 EUR
ORIGCF503101
Anti-SMS Mouse Monoclonal Antibody [clone: 2H1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SMS
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR