Anti-LSM1 Mouse Monoclonal Antibody [clone: 9D7]
Toimittaja: OriGene
ORIGCF503106EA 749 EUR
ORIGCF503106
Anti-LSM1 Mouse Monoclonal Antibody [clone: 9D7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LSM1
Clonality: Monoclonal
Clone: 9D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human