Anti-RHOJ Mouse Monoclonal Antibody [clone: 1C8]

Toimittaja: OriGene
ORIGCF505499EA 749 EUR
ORIGCF505499
Anti-RHOJ Mouse Monoclonal Antibody [clone: 1C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RHOJ
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR