Anti-PPARA Mouse Monoclonal Antibody [clone: 2D10]

Toimittaja: OriGene
ORIGCF503529EA 749 EUR
ORIGCF503529
Anti-PPARA Mouse Monoclonal Antibody [clone: 2D10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPARA
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR