Anti-DAND5 Mouse Monoclonal Antibody [clone: 2H3]

Toimittaja: OriGene
ORIGCF503555EA 749 EUR
ORIGCF503555
Anti-DAND5 Mouse Monoclonal Antibody [clone: 2H3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DAND5
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR