Anti-CAPN9 Mouse Monoclonal Antibody [clone: 1C2]

Toimittaja: OriGene
ORIGCF503569EA 749 EUR
ORIGCF503569
Anti-CAPN9 Mouse Monoclonal Antibody [clone: 1C2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CAPN9
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR